Assay Details
Target Gene Details
Entrez Gene ID: | 71661 |
Gene Name: | RIKEN cDNA 0610005C13 gene |
Gene Aliases: |
AI182092 |
Location: |
Chr.7:45567795-45575176 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 0610005C13Rik | NR_038165.1 | |||
| NR_038166.1 | ||||
| AK002217.1 | ||||
| AK085337.1 | 4 | 1150 |
Target Gene Details
Entrez Gene ID: | 12036 |
Gene Name: | branched chain aminotransferase 2, mitochondrial |
Gene Aliases: |
Bcat-2, Eca40 |
Location: |
Chr.7:45569855-45589710 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcat2 | NM_009737.3 | NP_033867.1 | ||
| AF031467.1 | AAC96067.1 | |||
| AK037482.1 | BAC29815.1 | |||
| AK145300.1 | BAE26355.1 | |||
| BB630667.1 | ||||
| BC017688.1 | AAH17688.1 | |||
| BC048072.1 | AAH48072.1 | |||
| BY279029.1 | ||||
| U68526.1 | AAB67674.1 |
More Information
Set Membership: |
|

Genomic Map