Assay Details
Target Gene Details
Entrez Gene ID: | 12036 |
Gene Name: | branched chain aminotransferase 2, mitochondrial |
Gene Aliases: |
Bcat-2, Eca40 |
Location: |
Chr.7:45569855-45589710 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcat2 | NM_001243052.1 | 6 | 824 | NP_001229981.1 |
| NM_001243053.1 | 7 | 709 | NP_001229982.1 | |
| NM_009737.3 | 6 | 797 | NP_033867.1 | |
| AF031467.1 | 6 | 589 | AAC96067.1 | |
| AK037482.1 | 6 | 602 | BAC29815.1 | |
| AK137049.1 | 7 | 760 | BAE23217.1 | |
| AK145300.1 | 6 | 642 | BAE26355.1 | |
| AK160159.1 | 6 | 825 | BAE35664.1 | |
| AK192808.1 | ||||
| BB630667.1 | 6 | 600 | ||
| BC017688.1 | 6 | 574 | AAH17688.1 | |
| BC048072.1 | 6 | 588 | AAH48072.1 | |
| U68526.1 | 6 | 583 | AAB67674.1 |
More Information
Set Membership: |
|

Genomic Map