Assay Details
Target Gene Details
Entrez Gene ID: | 12036 |
Gene Name: | branched chain aminotransferase 2, mitochondrial |
Gene Aliases: |
Bcat-2, Eca40 |
Location: |
Chr.7:45569855-45589710 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcat2 | NM_001243052.1 | NP_001229981.1 | ||
| NM_001243053.1 | NP_001229982.1 | |||
| NM_009737.3 | NP_033867.1 | |||
| AF031467.1 | AAC96067.1 | |||
| AK137049.1 | BAE23217.1 | |||
| AK145300.1 | BAE26355.1 | |||
| AK160159.1 | BAE35664.1 | |||
| BC017688.1 | AAH17688.1 | |||
| BC048072.1 | AAH48072.1 |
More Information
Set Membership: |
|

Genomic Map