Assay Details
Target Gene Details
Entrez Gene ID: | 11812 |
Gene Name: | apolipoprotein C-I |
Gene Aliases: |
Apo-CIB, ApoC-IB, apo-CI, apoC-I |
Location: |
Chr.7:19689480-19692659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apoc1 | NM_001110009.2 | NP_001103479.1 | ||
| NM_007469.5 | NP_031495.1 | |||
| AK011358.1 | BAB27566.1 | |||
| BC019398.1 | AAH19398.1 | |||
| BC094638.1 | AAH94638.1 | |||
| BY087738.1 | ||||
| CB947285.1 |
More Information
Set Membership: |
|

Genomic Map