Assay Details
Target Gene Details
Entrez Gene ID: | 11425 |
Gene Name: | apolipoprotein C-IV |
Gene Aliases: |
Acl |
Location: |
Chr.7:19678084-19681460 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apoc4 | NM_007385.3 | 3 | 444 | NP_031411.1 |
| BC024657.1 | AAH24657.1 |
More Information
Set Membership: |
|

Genomic Map