Assay Details
Target Gene Details
Entrez Gene ID: | 71241 |
Gene Name: | doublesex and mab-3 related transcription factor like family C2 |
Gene Aliases: |
4933432E21Rik, Dmrt7 |
Location: |
Chr.7:24869876-24877655 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmrtc2 | NM_027732.2 | NP_082008.1 | ||
| AF542046.1 | AAN77233.1 | |||
| AK017020.1 | ||||
| BC100434.1 | AAI00435.1 | |||
| BC138203.1 | ||||
| BC145988.1 |
More Information
Set Membership: |
|

Genomic Map