Assay Details
Target Gene Details
Entrez Gene ID: | 245128 |
Gene Name: | expressed sequence AU018091 |
Gene Aliases: |
- |
Location: |
Chr.7:3154658-3169252 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AU018091 | NM_001004153.2 | 10 | 1813 | NP_001004153.2 |
| AB201954.1 | 10 | 1764 | BAF02914.1 | |
| BC050126.1 | 10 | 1692 | AAH50126.2 | |
| BC080735.1 | 10 | 1821 | AAH80735.1 | |
| BC086454.1 | 8 | 1737 | AAH86454.1 |
More Information
Set Membership: |
|

Genomic Map