Assay Details
Target Gene Details
Entrez Gene ID: | 245128 |
Gene Name: | expressed sequence AU018091 |
Gene Aliases: |
- |
Location: |
Chr.7:3154658-3169252 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AU018091 | NM_001004153.2 | NP_001004153.2 | ||
| AB201954.1 | BAF02914.1 | |||
| BC050126.1 | AAH50126.2 | |||
| BC080735.1 | AAH80735.1 | |||
| BC086454.1 | AAH86454.1 |
More Information
Set Membership: |
|

Genomic Map