Assay Details
Target Gene Details
Entrez Gene ID: | 233879 |
Gene Name: | aspartate beta-hydroxylase domain containing 1 |
Gene Aliases: |
A830007L07Rik, Gm168 |
Location: |
Chr.7:126945981-126949886 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asphd1 | NM_001039645.1 | NP_001034734.1 | ||
| AK043553.1 | ||||
| BC111101.1 | AAI11102.1 |
More Information
Set Membership: |
|

Genomic Map