Assay Details
Target Gene Details
Entrez Gene ID: | 320871 |
Gene Name: | RIKEN cDNA B230206H07 gene |
Gene Aliases: |
- |
Location: |
Chr.7:141359178-141365110 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B230206H07Rik | NR_033532.1 | |||
| AK038776.1 | BAC30130.1 | |||
| AK080797.1 |

Genomic Map