Assay Details
Target Gene Details
Entrez Gene ID: | 12348 |
Gene Name: | carbonic anhydrase 11 |
Gene Aliases: |
Ca11 |
Location: |
Chr.7:45699848-45704671 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car11 | NM_009800.4 | NP_033930.1 | ||
| AB079601.1 | BAC07262.1 | |||
| AK013465.1 | ||||
| AK140073.1 | BAE24229.1 | |||
| AY075026.1 | AAL78174.1 | |||
| BC019393.1 | AAH19393.1 |

Genomic Map