Assay Details
Target Gene Details
Entrez Gene ID: | 14256 |
Gene Name: | FMS-like tyrosine kinase 3 ligand |
Gene Aliases: |
Flt3lg, Ly72L |
Location: |
Chr.7:45131184-45136490 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flt3l | NM_013520.3 | NP_038548.3 | ||
| NR_102725.1 | ||||
| AK020105.1 | BAB31997.1 |
More Information
Set Membership: |
|

Genomic Map