Assay Details
Target Gene Details
Entrez Gene ID: | 19144 |
Gene Name: | kallikrein related-peptidase 6 |
Gene Aliases: |
AI849898, BSP, Bssp, Klk29, MSP, Prss18, Prss9, neurosin |
Location: |
Chr.7:43824544-43832027 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klk6 | NM_001164696.1 | 5 | 583 | NP_001158168.1 |
| NM_001164697.1 | 5 | 579 | NP_001158169.1 | |
| NM_001164698.1 | 5 | 480 | NP_001158170.1 | |
| NM_011177.2 | 4 | 591 | NP_035307.2 | |
| AB008928.1 | 5 | 569 | BAB55605.1 | |
| AB015206.1 | 5 | 567 | BAA28895.1 | |
| AK138218.1 | 5 | 580 | BAE23586.1 | |
| BC031119.1 | 4 | 428 | AAH31119.1 | |
| EL956218.1 | ||||
| Y18723.1 | 4 | 591 | CAA77269.1 |
More Information
Set Membership: |
|

Genomic Map