Assay Details
Target Gene Details
Entrez Gene ID: | 56209 |
Gene Name: | glycerophosphodiester phosphodiesterase 1 |
Gene Aliases: |
1200003M13Rik, Mir16, RGS16 |
Location: |
Chr.7:118688558-118705738 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gde1 | NM_019580.4 | 3 | 578 | NP_062526.1 |
| AF212860.1 | 3 | 559 | AAF65232.1 | |
| AK004583.1 | 3 | 591 | ||
| AK005361.1 | 3 | 579 | BAB23975.1 | |
| AK150807.1 | 3 | 600 | BAE29870.1 | |
| BC003902.1 | 3 | 537 | AAH03902.1 |
More Information
Set Membership: |
|

Genomic Map