Assay Details
Target Gene Details
Entrez Gene ID: | 68616 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 3 |
Gene Aliases: |
1110015E22Rik |
Location: |
Chr.7:126766414-126775645 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdpd3 | NM_024228.2 | NP_077190.2 | ||
| AK003726.1 | BAB22961.1 |
More Information
Set Membership: |
|

Genomic Map