Assay Details
Target Gene Details
Entrez Gene ID: | 68695 |
Gene Name: | HD domain containing 3 |
Gene Aliases: |
1110033O09Rik, C86475, MESH1 |
Location: |
Chr.7:80343083-80346097 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hddc3 | NM_026812.2 | NP_081088.1 | ||
| AK004078.1 | BAB23158.1 | |||
| BC038310.1 | AAH38310.1 |
More Information
Set Membership: |
|

Genomic Map