Assay Details
Target Gene Details
Entrez Gene ID: | 622320 |
Gene Name: | potassium channel tetramerisation domain containing 21 |
Gene Aliases: |
EG622320 |
Location: |
Chr.7:97332311-97350216 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kctd21 | NM_001039039.3 | NP_001034128.2 | ||
| AK141547.1 | BAE24730.1 | |||
| BB852194.1 |
Target Gene Details
Entrez Gene ID: | 244144 |
Gene Name: | ubiquitin specific peptidase 35 |
Gene Aliases: |
Gm1088, Gm493 |
Location: |
Chr.7:97309380-97332255 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp35 |
More Information
Set Membership: |
|

Genomic Map