Assay Details
Target Gene Details
Entrez Gene ID: | 14132 |
Gene Name: | Fc receptor, IgG, alpha chain transporter |
Gene Aliases: |
FcRn |
Location: |
Chr.7:45092992-45103846 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcgrt | NM_010189.3 | NP_034319.2 | ||
| AK146933.1 | ||||
| BC003786.1 | AAH03786.1 | |||
| D37874.1 | BAA07111.1 | |||
| L17022.1 | AAA16904.1 |
More Information
Set Membership: |
|

Genomic Map