Assay Details
Target Gene Details
Entrez Gene ID: | 70300 |
Gene Name: | fuzzy homolog (Drosophila) |
Gene Aliases: |
2600013E07Rik, b2b1273Clo |
Location: |
Chr.7:44895265-44900624 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fuz | NM_027376.3 | NP_081652.2 | ||
| AK033247.1 | ||||
| AK134650.1 | BAE22226.1 | |||
| AK168939.1 | ||||
| AK188272.1 | ||||
| BC045601.1 | 5 | 1549 | AAH45601.1 | |
| BC057127.1 | 1 | 256 | AAH57127.1 | |
| CO427180.1 |

Genomic Map