Assay Details
Target Gene Details
Entrez Gene ID: | 101502 |
Gene Name: | hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7 |
Gene Aliases: |
AI195443, BB098564 |
Location: |
Chr.7:127800609-127803802 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsd3b7 | NM_001040684.1 | NP_001035774.1 | ||
| NM_133943.2 | NP_598704.2 | |||
| AF277718.1 | AAG37823.1 | |||
| AK077476.1 | ||||
| BB854484.1 | ||||
| BC132605.1 | ||||
| BC138589.1 | ||||
| BY133253.1 |
More Information
Set Membership: |
|

Genomic Map