Assay Details
Target Gene Details
Entrez Gene ID: | 70973 |
Gene Name: | RIKEN cDNA 4931431B13 gene |
Gene Aliases: |
1700108I23Rik, AA536668 |
Location: |
Chr.7:128192050-128203328 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931431B13Rik | NR_045183.1 | |||
| AK007137.1 |
Target Gene Details
Entrez Gene ID: | 381924 |
Gene Name: | integrin, alpha D |
Gene Aliases: |
Cd11d |
Location: |
Chr.7:128173933-128205521 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgad | NM_001029872.3 | 21 | 2579 | NP_001025043.3 |
| AK156218.1 | 21 | 2580 | BAE33629.1 |
More Information
Set Membership: |
|

Genomic Map