Assay Details
Target Gene Details
Entrez Gene ID: | 53415 |
Gene Name: | HIV-1 tat interactive protein 2, homolog (human) |
Gene Aliases: |
AW111545, Cc3, Tip30 |
Location: |
Chr.7:49759045-49778364 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htatip2 | NM_001146049.1 | 1 | 63 | NP_001139521.1 |
| NM_001146050.1 | NP_001139522.1 | |||
| NM_001146053.1 | NP_001139525.1 | |||
| NM_016865.3 | 1 | 63 | NP_058561.2 | |
| AK015389.1 | BAB29825.1 | |||
| AK144837.1 | BAE26091.1 | |||
| AK146400.1 | BAE27141.1 | |||
| AK151254.1 | BAE30243.1 | |||
| AK152418.1 | BAE31203.1 | |||
| BB575839.2 | ||||
| BC004083.1 | AAH04083.1 | |||
| BY292123.1 | 1 | 65 | ||
| CB593552.1 |
More Information
Set Membership: |
|

Genomic Map