Assay Details
Target Gene Details
Entrez Gene ID: | 269951 |
Gene Name: | isocitrate dehydrogenase 2 (NADP+), mitochondrial |
Gene Aliases: |
E430004F23, IDPm, Idh-2 |
Location: |
Chr.7:80094846-80115387 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Idh2 | NM_173011.2 | 10 | 1303 | NP_766599.2 |
| AF212319.1 | 10 | 1274 | AAG43538.1 | |
| AK088110.1 | 10 | 1275 | BAC40149.1 | |
| AK145753.1 | 10 | 1248 | BAE26628.1 | |
| AK146564.1 | 10 | 1389 | ||
| AK161640.1 | 10 | 1287 | BAE36505.1 | |
| AK169395.1 | 10 | 1300 | BAE41142.1 | |
| AK179419.1 | ||||
| AK209654.1 | ||||
| AK212737.1 | ||||
| BC060030.1 | 10 | 1291 | AAH60030.1 | |
| BQ747924.1 | 1 | 52 |
More Information
Set Membership: |
|

Genomic Map