Assay Details
Target Gene Details
Entrez Gene ID: | 269951 |
Gene Name: | isocitrate dehydrogenase 2 (NADP+), mitochondrial |
Gene Aliases: |
E430004F23, IDPm, Idh-2 |
Location: |
Chr.7:80094846-80115387 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Idh2 | NM_173011.2 | 8 | 1101 | NP_766599.2 |
| AF212319.1 | 8 | 1072 | AAG43538.1 | |
| AK088110.1 | 8 | 1073 | BAC40149.1 | |
| AK145753.1 | 8 | 1046 | BAE26628.1 | |
| AK146564.1 | 8 | 1187 | ||
| AK161640.1 | 8 | 1085 | BAE36505.1 | |
| AK169395.1 | 8 | 1098 | BAE41142.1 | |
| AK177496.1 | ||||
| BC060030.1 | 8 | 1089 | AAH60030.1 |
More Information
Set Membership: |
|

Genomic Map