Assay Details
Target Gene Details
Entrez Gene ID: | 16866 |
Gene Name: | luteinizing hormone beta |
Gene Aliases: |
LH, LH-B, LSH-B, LSH-beta |
Location: |
Chr.7:45418166-45421855 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lhb | NM_008497.2 | NP_032523.2 | ||
| BB618381.1 |
More Information
Set Membership: |
|

Genomic Map