Assay Details
Target Gene Details
Entrez Gene ID: | 317652 |
Gene Name: | kallikrein related-peptidase 15 |
Gene Aliases: |
- |
Location: |
Chr.7:43929405-43939852 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klk15 | NM_174865.1 | NP_777354.1 | ||
| BC106965.1 | AAI06966.1 | |||
| BC106966.2 | AAI06967.1 | |||
| BC107012.1 | AAI07013.1 |
More Information
Set Membership: |
|

Genomic Map