Assay Details
Target Gene Details
Entrez Gene ID: | 101613 |
Gene Name: | NLR family, pyrin domain containing 6 |
Gene Aliases: |
AI504961, Avr, Nalp6, Navr, Navr/Avr, Pypaf5 |
Location: |
Chr.7:140920902-140929192 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp6 | NM_133946.2 | NP_598707.2 | ||
| FJ222825.1 | ||||
| JF810536.1 |
More Information
Set Membership: |
|

Genomic Map