Assay Details
Target Gene Details
Entrez Gene ID: | 16866 |
Gene Name: | luteinizing hormone beta |
Gene Aliases: |
LH, LH-B, LSH-B, LSH-beta |
Location: |
Chr.7:45418166-45421855 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lhb | NM_008497.2 | NP_032523.2 | ||
| AV173269.2 | ||||
| BB618381.1 | ||||
| BM224915.2 |
Target Gene Details
Entrez Gene ID: | 20174 |
Gene Name: | RuvB-like protein 2 |
Gene Aliases: |
mp47, p47 |
Location: |
Chr.7:45421896-45435170 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ruvbl2 | NM_011304.3 | NP_035434.1 | ||
| AK135404.1 | BAE22521.1 | |||
| AK144156.1 | BAE25736.1 | |||
| AK159112.1 | BAE34828.1 | |||
| AK186221.1 | ||||
| AK188084.1 | 2 | 190 | ||
| AK219298.1 |
More Information
Set Membership: |
|

Genomic Map