Assay Details
Target Gene Details
Entrez Gene ID: | 211548 |
Gene Name: | nodal modulator 1 |
Gene Aliases: |
D7Ertd156e, Nomo, pM5 |
Location: |
Chr.7:46033696-46084212 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nomo1 | NM_153057.4 | NP_694697.3 | ||
| AK030584.1 | ||||
| AK035957.1 | ||||
| AK080754.1 | ||||
| AK082495.1 | BAC38508.1 | |||
| AK082963.1 | BAC38713.1 | |||
| AK142338.1 | ||||
| AK150107.1 | ||||
| AK165643.1 | ||||
| AK166764.1 | BAE39002.1 | |||
| BC033923.1 | AAH33923.1 | |||
| BC072630.1 | AAH72630.1 |

Genomic Map