Assay Details
Target Gene Details
Entrez Gene ID: | 211548 |
Gene Name: | nodal modulator 1 |
Gene Aliases: |
D7Ertd156e, Nomo, pM5 |
Location: |
Chr.7:46033696-46084212 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nomo1 | NM_153057.4 | 21 | 2545 | NP_694697.3 |
| AK030584.1 | 21 | 2512 | ||
| AK035957.1 | 21 | 2517 | ||
| AK080754.1 | 21 | 2408 | ||
| AK082495.1 | 21 | 2424 | BAC38508.1 | |
| AK082963.1 | 21 | 2404 | BAC38713.1 | |
| AK142338.1 | 21 | 2515 | ||
| AK150107.1 | 21 | 2500 | ||
| AK165643.1 | 21 | 2408 | ||
| AK166764.1 | 6 | 555 | BAE39002.1 | |
| AK190236.1 | ||||
| AK200826.1 | ||||
| AK201878.1 | ||||
| AK201938.1 | ||||
| AK202854.1 | ||||
| AK203657.1 | ||||
| AK205783.1 | ||||
| AK211946.1 | ||||
| AK213173.1 | 2 | 162 | ||
| BC033923.1 | 10 | 1129 | AAH33923.1 | |
| BC072630.1 | 21 | 2491 | AAH72630.1 |

Genomic Map