Assay Details
Target Gene Details
Entrez Gene ID: | 84113 |
Gene Name: | prostate tumor over expressed gene 1 |
Gene Aliases: |
1110030G05Rik, AU041779, Gcap3 |
Location: |
Chr.7:44863067-44870411 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptov1 | NM_133949.1 | NP_598710.1 | ||
| AK003974.1 | BAB23101.1 | |||
| AK187740.1 | ||||
| AK219728.1 | ||||
| BC008543.1 | AAH08543.1 | |||
| BC024632.1 | AAH24632.1 |
More Information
Set Membership: |
|

Genomic Map