Assay Details
Target Gene Details
Entrez Gene ID: | 17188 |
Gene Name: | MYC-associated zinc finger protein (purine-binding transcription factor) |
Gene Aliases: |
PUR1, Pur-1, SAF-1, SAF-2 |
Location: |
Chr.7:127022123-127026553 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Maz | NM_010772.1 | 4 | 1208 | NP_034902.1 |
| AK209702.1 | 2 | 59 | ||
| BC004758.1 | 6 | 745 | ||
| DQ150586.1 | 3 | 922 | ||
| L04649.1 | 4 | 1181 |
More Information
Set Membership: |
|

Genomic Map