Assay Details
Target Gene Details
Entrez Gene ID: | 66091 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3 |
Gene Aliases: |
1010001M12Rik, 1700022J01Rik |
Location: |
Chr.7:3617373-3620161 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufa3 | NM_025348.2 | NP_079624.1 | ||
| AK003133.1 | BAB22593.1 | |||
| AK006243.1 | BAB24477.1 | |||
| AK019099.1 | BAB31545.1 | |||
| BC120712.1 | ||||
| BC120714.1 |
More Information
Set Membership: |
|

Genomic Map