Assay Details
Target Gene Details
Entrez Gene ID: | 102466795 |
Gene Name: | microRNA 7049 |
Gene Aliases: |
Gm27277, mmu-mir-7049 |
Location: |
Chr.7:28560925-28560984 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir7049 | NR_106016.1 |
Target Gene Details
Entrez Gene ID: | 70584 |
Gene Name: | p21 protein (Cdc42/Rac)-activated kinase 4 |
Gene Aliases: |
5730488L07Rik, AW555722, mKIAA1142 |
Location: |
Chr.7:28558819-28598186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pak4 | NM_027470.3 | NP_081746.1 | ||
| AK017713.1 | BAB30889.1 | |||
| AK088512.1 | BAC40396.1 | |||
| AK129298.2 | BAC98108.2 | |||
| AK183660.1 | ||||
| AY217016.1 | AAO61496.1 | |||
| BC030389.1 | AAH30389.1 | |||
| BC048238.1 | AAH48238.1 |
More Information
Set Membership: |
|

Genomic Map