Assay Details
Target Gene Details
Entrez Gene ID: | 72981 |
Gene Name: | protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor) |
Gene Aliases: |
2900052B10Rik, Dap4, Rpkrir |
Location: |
Chr.7:98703230-98718062 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkrir | NM_028410.1 | NP_082686.1 | ||
| AK013663.1 | BAB28945.1 | |||
| BC049103.1 | AAH49103.1 |

Genomic Map