Assay Details
Target Gene Details
Entrez Gene ID: | 83485 |
Gene Name: | neugrin, neurite outgrowth associated |
Gene Aliases: |
AW552001 |
Location: |
Chr.7:80261215-80265378 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ngrn | NM_031375.4 | NP_113552.3 | ||
| NR_028053.1 | ||||
| AB047544.1 | BAB21528.1 | |||
| AK083330.1 | BAC38868.1 | |||
| AK144756.1 | BAE26050.1 | |||
| AK146059.1 | BAE26868.1 | |||
| AK195192.1 | ||||
| CJ154012.1 |

Genomic Map