Assay Details
Target Gene Details
Entrez Gene ID: | 67369 |
Gene Name: | glutaminyl-peptide cyclotransferase-like |
Gene Aliases: |
1810019P04Rik, BB101812, gQC |
Location: |
Chr.7:19140217-19149196 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qpctl | NM_026111.3 | NP_080387.2 | ||
| AK007555.1 | BAB25105.1 | |||
| AK007701.1 | BAB25199.1 | |||
| AK035587.1 | BAC29114.1 | |||
| AK045687.1 | BAC32458.1 | |||
| AK148755.1 | 2 | 951 | ||
| BC058181.1 | AAH58181.1 |
More Information
Set Membership: |
|

Genomic Map