Assay Details
Target Gene Details
Entrez Gene ID: | 67773 |
Gene Name: | K(lysine) acetyltransferase 8 |
Gene Aliases: |
2010203C02Rik, 5830450F21Rik, D7Ertd629e, MOF, Myst1 |
Location: |
Chr.7:127912517-127925763 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat8 | NM_026370.1 | 3 | 379 | NP_080646.1 |
| AK003264.1 | 3 | 379 | BAB22680.1 | |
| AK008418.1 | 3 | 373 | ||
| AK017161.1 | 2 | 125 | ||
| AK018002.1 | 3 | 376 | BAC25539.1 | |
| AK029350.1 | 3 | 381 | BAC26411.1 | |
| AK132000.1 | 3 | 380 | BAE20930.1 | |
| AK146710.1 | 3 | 381 | BAE27376.1 | |
| BC036284.1 | 3 | 387 | AAH36284.1 |
More Information
Set Membership: |
|

Genomic Map