Assay Details
Target Gene Details
Entrez Gene ID: | 272350 |
Gene Name: | predicted gene 5065 |
Gene Aliases: |
EG272350 |
Location: |
Chr.7:5350542-5360682 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5065 | NR_003622.2 |
Target Gene Details
Entrez Gene ID: | 232827 |
Gene Name: | NLR family, pyrin domain containing 2 |
Gene Aliases: |
E330007A02Rik, NBS1, Nalp2, PAN1, PYPAF2 |
Location: |
Chr.7:5298547-5351035 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp2 | NM_177690.3 | NP_808358.2 | ||
| AK054264.1 | BAC35710.1 | |||
| AY360473.1 | AAQ64011.1 | |||
| DQ066881.1 | AAY68479.1 |
More Information
Set Membership: |
|

Genomic Map