Assay Details
Target Gene Details
Entrez Gene ID: | 243967 |
Gene Name: | netrin 5 |
Gene Aliases: |
Gm484 |
Location: |
Chr.7:45684022-45694556 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntn5 | NM_001033356.3 | 1 | 394 | NP_001028528.2 |
| NM_001289692.1 | 1 | 394 | NP_001276621.1 | |
| NM_001289693.1 | 1 | 394 | NP_001276622.1 | |
| AK042348.1 | 1 | 395 | ||
| AK142898.1 | BAE25222.1 | |||
| BC150901.1 |
Target Gene Details
Entrez Gene ID: | 56546 |
Gene Name: | secretory blood group 1 |
Gene Aliases: |
Fut10, Fut3 |
Location: |
Chr.7:45677686-45694402 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec1 | NM_001271578.1 | NP_001258507.1 | ||
| AK030246.1 | BAC26862.1 |
More Information
Set Membership: |
|

Genomic Map