Assay Details
Target Gene Details
Entrez Gene ID: | 56330 |
Gene Name: | programmed cell death 5 |
Gene Aliases: |
2200003D22Rik, AA408513, Tfar19 |
Location: |
Chr.7:35641985-35647482 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdcd5 | NM_019746.4 | NP_062720.1 | ||
| AF161074.1 | AAD45607.1 | |||
| AK007297.1 | BAB24940.1 | |||
| AK008632.1 | BAB25794.1 | |||
| BC048476.1 | AAH48476.1 | |||
| BC056167.1 | AAH56167.1 | |||
| BC092092.1 | AAH92092.1 |
More Information
Set Membership: |
|

Genomic Map