Assay Details
Target Gene Details
Entrez Gene ID: | 70314 |
Gene Name: | rabaptin, RAB GTPase binding effector protein 2 |
Gene Aliases: |
2610011A08Rik, AI462746, Fra |
Location: |
Chr.7:126428416-126445907 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabep2 | NM_030566.2 | NP_085043.2 | ||
| AK089938.1 | ||||
| AK144836.1 | BAE26090.1 | |||
| AK157832.1 | BAE34216.1 | |||
| AK206828.1 | ||||
| BC004088.1 | 1 | 158 | AAH04088.1 | |
| BU514356.1 |

Genomic Map