Assay Details
Target Gene Details
Entrez Gene ID: | 69358 |
Gene Name: | leucine rich repeat containing 51 |
Gene Aliases: |
1700008D07Rik, Lrtomt |
Location: |
Chr.7:101912989-101933917 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc51 | NM_001162973.1 | NP_001156445.1 | ||
| NM_027053.1 | NP_081329.1 | |||
| AK005758.1 | BAB24223.1 |

Genomic Map