Assay Details
Target Gene Details
Entrez Gene ID: | 69358 |
Gene Name: | leucine rich repeat containing 51 |
Gene Aliases: |
1700008D07Rik, Lrtomt |
Location: |
Chr.7:101912989-101933917 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc51 | NM_001162973.1 | 2 | 170 | NP_001156445.1 |
| NM_001162974.1 | 1 | 76 | NP_001156446.1 | |
| NM_027053.1 | 3 | 233 | NP_081329.1 | |
| AK005758.1 | 3 | 184 | BAB24223.1 | |
| BC025128.1 | 1 | 127 | AAH25128.1 | |
| EU627089.1 | ||||
| EU627090.1 | 1 | 76 |

Genomic Map