Assay Details
Target Gene Details
Entrez Gene ID: | 18631 |
Gene Name: | peroxisomal biogenesis factor 11 alpha |
Gene Aliases: |
PEX11alpha |
Location: |
Chr.7:79735957-79743126 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pex11a | AK076776.1 |
Target Gene Details
Entrez Gene ID: | 626359 |
Gene Name: | WD repeat domain 93 |
Gene Aliases: |
EG626359 |
Location: |
Chr.7:79743135-79785952 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdr93 | NM_001037927.1 | NP_001033016.1 | ||
| AB205021.1 | BAE19974.1 |
More Information
Set Membership: |
|

Genomic Map