Assay Details
Target Gene Details
Entrez Gene ID: | 24075 |
Gene Name: | TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor |
Gene Aliases: |
30kDa, AU041226, TAFII30, Taf2h |
Location: |
Chr.7:105742894-105744375 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf10 | NM_020024.3 | NP_064408.2 | ||
| BC031418.1 | AAH31418.1 | |||
| BC080311.1 | AAH80311.1 |
More Information
Set Membership: |
|

Genomic Map