Assay Details
Target Gene Details
Entrez Gene ID: | 353499 |
Gene Name: | transmembrane channel-like gene family 4 |
Gene Aliases: |
- |
Location: |
Chr.7:3665754-3677553 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmc4 | NM_181820.2 | NP_861541.2 | ||
| AK050105.1 | BAC34068.1 | |||
| AY236493.1 | AAP69871.1 | |||
| AY263162.1 | AAP78777.1 | |||
| BC022758.1 | AAH22758.1 | |||
| BC040466.1 | AAH40466.1 | |||
| CN719678.1 |

Genomic Map