Assay Details
Target Gene Details
Entrez Gene ID: | 74237 |
Gene Name: | tubulin, gamma complex associated protein 2 |
Gene Aliases: |
1700022B05Rik |
Location: |
Chr.7:139995955-140036674 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubgcp2 | NM_001286007.1 | 1 | 171 | NP_001272936.1 |
| NM_001286009.1 | 1 | 171 | NP_001272938.1 | |
| BY234488.1 | 1 | 175 |
Target Gene Details
Entrez Gene ID: | 69752 |
Gene Name: | zinc finger protein 511 |
Gene Aliases: |
2410004P19Rik, C86579, Znf511 |
Location: |
Chr.7:140036391-140040606 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp511 | NM_027201.1 | 1 | 114 | NP_081477.1 |
| AK010407.1 | 1 | 102 | BAB26915.1 |
More Information
Set Membership: |
|

Genomic Map