Assay Details
Target Gene Details
Entrez Gene ID: | 69752 |
Gene Name: | zinc finger protein 511 |
Gene Aliases: |
2410004P19Rik, C86579, Znf511 |
Location: |
Chr.7:140036391-140040606 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp511 | NM_027201.1 | NP_081477.1 | ||
| AK010407.1 | BAB26915.1 | |||
| BC065404.1 | AAH65404.1 |

Genomic Map