Assay Details
Target Gene Details
Entrez Gene ID: | 22282 |
Gene Name: | upstream transcription factor 2 |
Gene Aliases: |
Usf-2, bHLHb12 |
Location: |
Chr.7:30945248-30957250 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usf2 | NM_011680.2 | 4 | 418 | NP_035810.1 |
| AB221562.1 | 4 | 377 | ||
| AK146871.1 | 4 | 365 | BAE27494.1 | |
| AK158906.1 | ||||
| AK211311.1 | 1 | 50 | ||
| BC082995.1 | 4 | 418 | AAH82995.1 | |
| U01662.1 | 4 | 434 | AAA20492.1 | |
| U01663.1 | AAA20493.1 |

Genomic Map